Learn More
R&D Systems™ Reconstitution Buffer 5 (Acetic Acid)
Codice prodotto. 30001090
Click to view available options
Quantità:
2 mL
Dimensione della confezione:
2mL
Codice prodotto. 30001090
Marca: R&D Systems™ RB05
100 mM Acetic Acid
This product is intended as a companion product for reconstitution of selected lyophilized antibody or protein to maximize recovery and activity. Refer to the specific antibody or protein product data sheet for the recommended reconstitution concentrations.
TRUSTED_SUSTAINABILITY
Specifica
Quantità 2 mL