Learn More
Eppendorf™ Reference™ 2 BASIC - Trade-In Pipette
GREENER_CHOICE
Codice prodotto. 15773489 Sfoglia Tutto Eppendorf Prodotti
Click to view available options
Volume (sistema metrico):
0.1 to 2.5 μL
0.25 to 2.5 mL
0.5 to 10 μL
0.5 to 5 mL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Colore:
Blue
Dark Gray
Light Gray
Medium Gray
Orange
Red
Turquoise
Violet
Yellow
Dimensione della confezione:
Pezzo
Codice prodotto. 15773489
Marca: Eppendorf™ 4924000240
Minimize risk of aerosol contamination and reduce thumb strain with volume pipettes
  • Integrated pipetting system with internal RFID technology
  • Serial number is printed on multiple components of pipette
  • Color coded and volume labeled pipette for quick identification of volume size/tip size
  • Round upper part of pipette makes it possible to work in every position
  • Four-digit display for exact, quick and secure volume setting (volume lock included)
  • High flexibility with ability to remove single cones
  • Ensures continuous identical pipet alignment during work process
  • Fully autoclavable pipet with smooth surface and few rough edges
  • Ergonomic handling with reduced operating effort
  • Simple and quick tip ejection with active aerosol reduction
  • Reduce repetitive strain injuries by not having to move your thumb sideways
  • High precision and accuracy provides reliable pipetting results
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità Lot Specific Certificate
Punte compatibili epT.I.P.S.
Spaziatura punta regolabile No
Autoclavabile Autoclavable
Colore Yellow
Monouso Reusable
Da utilizzare con epT.I.P.S. 200 μL
Tipo di impugnatura Ergonomic
Materiale Plastic
Numero di canali 1
Programmabilità No
Sterilità Non-sterile
Tipo Manual Pipette
Volume (sistema metrico) 2 to 20 μL
Da utilizzare con (applicazione) Pipette Tips
Da utilizzare con (apparecchiatura) Pipette Tips