Learn More
Cole-Parmer Reinforced Thermoplastic Elastomer (TPE) Tubing
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1 in.
1/2 in.
1/4 in.
1/8 in.
3/4 in.
3/8 in.
5/8 in.
Diametro (sistema imperiale) esterno:
1 in.
1-1/8 in.
1-3/8 in.
1/2 in.
3/4 in.
3/8 in.
5/8 in.
Overall Length (English):
15 ft.
25 ft.
50 ft.
Dimensione della confezione:
15m
4.50m
7.60m
Descrizione
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. E in., cellent for use in peristaltic pump applications. Great alternative to silicone tubing. Does not contain PVC, DEHP, phthalates, or other plasticizers. Ultra-low e in., tractables and leachables. Low gas and o in., ygen permeability. Low protein binding and nonpyrogenic material. Thermally weldable. USP Class VI, USP 661, and RoHS compliant and BPOG tested
Specifica
Specifica
| Certificazione/Conformità | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diametro (sistema imperiale) interno | 5/8 in. |
| Materiale | TPE |
| Diametro (sistema imperiale) esterno | 1 in. |
| Overall Length (English) | 50 ft. |
| Metodo di sterilizzazione | E-beam/gamma irradiation between 25 and 45 kGy |
| Colore | Semitrasparente |