Learn More
Relative Density Standard 25°C, UKAS ISO 17025 / ISO 17034, 500 mL, nominal Relative Density 25/4 0.8693 at 25 °C, Paragon Scientific™
Click to view available options
Quantità:
500 mL
Dimensione della confezione:
500mL
Descrizione
- Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in accordance with primary level ASTM D1480 methodology
- The Relative density was calculated by dividing the density, as obtained from ASTM D1480, by the density of water at the reference temperature.
- The density of water was taken from the table denoted 'Density of Water' found in ASTM D1480 and confirms to the International Temperature Scale (ITS 90).
- Supplied in tamper evident packaging with a 2 year shelf life
- Manufactured in the United Kingdom
Specifica
Specifica
| Quantità | 500 mL |
Nominal values quoted. Certified Values will vary from batch to batch.