Learn More
Thermo Scientific™ Siero agglutinante Remel™, Salmonella H
Codice prodotto. 12759300
Click to view available options
Descrizione:
Polyvalent, Phase 2
Polyvalent, Phases 1 and 2
Rapid Diagnostic 1 (b, d, E, r)
Rapid Diagnostic 2 (b, E, k, L)
Rapid Diagnostic 3 (d, E, G, k)
Salmonella H (1, 2)
Salmonella H (1, 5)
Salmonella H (1, 6)
Salmonella H (1, 7)
Salmonella H (E) Polyvalent
Salmonella H (G) Polyvalent
Salmonella H (L) Polyvalent
Salmonella H (a)
Salmonella H (b)
Salmonella H (c )
Salmonella H (d)
Salmonella H (eh)
Salmonella H (enx)
Salmonella H (f, g)
Salmonella H (gm)
Salmonella H (i)
Salmonella H (r )
Salmonella H (y)
Salmonella H (z27)
Dimensione della confezione:
2mL
Pezzo
Codice prodotto. 12759300
Marca: Thermo Scientific™ R30162201
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY
Specifica
Set di controllo Positive
Descrizione Salmonella H (r )
Analiti rilevabili Antigeni H flagellari di ceppi di Salmonella per test di agglutinazione su vetrino e/o provetta
Tipo Agglutinating Sera
Tipo di imballaggio 2 mL/Vial
Quantità 2 mL
Target Organism Class Salmonella H