Learn More
Thermo Scientific™ Siero agglutinante Remel™, Salmonella O
Descrizione
Serologically identify somatic O antigens of Salmonella strains by slide agglutination test using Agglutinating Sera, Salmonella O.
Serologically identify somatic O antigens of Salmonella strains by slide agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O. The sera are absorbed to remove agglutinins for other Salmonella antigens and the paracolon α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O antigens.
- Select sera also for use as positive controls for stained Salmonella suspensions
Specifica
Specifica
| Set di controllo | Positive |
| Descrizione | Factor 10 (group E1) |
| Analiti rilevabili | Antigeni O somatici di ceppi di Salmonella per test di agglutinazione su vetrino e/o provetta |
| Tipo | Agglutinating Sera |
| Organism Group | Group E1 |
| Tipo di imballaggio | 2 mL/Vial |
| Quantità | 2 mL |
| Target Organism Class | Salmonella O |