Learn More
Renin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14291-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.Specifica
| Renin | |
| Polyclonal | |
| Unconjugated | |
| REN | |
| Angiotensinogenase, REN, Renin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19701, 24715, 5972 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P00797, P06281 | |
| REN, Ren1 | |
| Renin Fusion Protein Ag5612 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |