Learn More
Grant Instruments™ Replacement Basket for Grant Ultrasonic Baths
Descrizione
- Stainless steel construction
- Works in conjunction with lid to create a drip collection unit
- Prevents user from coming into contact with chemical solutions
- Prevents items from resting on the inside of the tank wall
- For use with Grant Ultrasonic Baths
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | XUB12 Ultrasonic Bath |
| Materiale | Stainless Steel |
| Size | (WxDxH) 263 x 203 x 193 mm |