Learn More
Grant Instruments™ Replacement Basket for Grant Ultrasonic Baths
Click to view available options
Da utilizzare con (apparecchiatura):
XUB12 Ultrasonic Bath
XUB18 Ultrasonic Bath
XUB25 Ultrasonic Bath
XUB5 Ultrasonic Bath
XUBA1 Ultrasonic Bath
XUBA3 Ultrasonic Bath
Dimensione della confezione:
Pezzo
Descrizione
- Stainless steel construction
- Works in conjunction with lid to create a drip collection unit
- Prevents user from coming into contact with chemical solutions
- Prevents items from resting on the inside of the tank wall
- For use with Grant Ultrasonic Baths
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | XUB25 Ultrasonic Bath |
| Materiale | Stainless Steel |
| Size | (WxDxH) 463 x 263 x 159 mm |