missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ resDNASEQ™ Quantitative CHO DNA Kits

Codice prodotto. 18634253 Sfoglia Tutto Applied Biosystems Prodotti
Cambia vista
Click to view available options
Include:
Assay Kit and Sample Preparation
Assay Kit only
Dimensione della confezione:
100 reazioni
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Include Dimensione della confezione
18634253 Assay Kit and Sample Preparation 100 reazioni
30000682 Assay Kit only Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18634253 Fornitore Applied Biosystems™ N. del fornitore 4413713
  • ONLINE EXCLUSIVE PRICE
    4769.78€
    4430.00€ / 100 reazioni
    Risparmia 339.78€
    7% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 16-10-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This quantitative PCR (qPCR)-based kit, part of the resDNASEQ™ Quantitative CHO DNA System, allows users to detect sub-picogram amounts of residual DNA from CHO cell line in under 5 hours.

This quantitative PCR (qPCR)-based kit, part of the resDNASEQ™ Quantitative CHO DNA System, enables you to detect sub-picogram amounts of residual DNA from the Chinese hamster ovary (CHO) cell line in under 5 hours. The system overcomes the limitations of traditional methods by combining high-recovery PrepSEQ™ sample preparation and TaqMan™-based quantitation of residual DNA.

Real-time quantitative PCR kit for highly sensitive residual CHO DNA quantitation
• Easy to use, results in under 5 hours
• Specific to CHO DNA, no cross-reactivity with unrelated DNA
• Part of an integrated system that includes sample preparation, TaqMan™ assay and master mix, standard DNA, instrument, and software
• Protocol available for automated sample preparation
• Flexible sample throughput allows you to perform rigorous DNA clearance studies

This kit is available with or without sample preparation reagents in the form of a PrepSEQ Residual DNA Sample Preparation Kit, which is optimized for highly efficient residual DNA recovery from complex mixtures of proteins, buffers, and salts.

Real-Time PCR for Highly Sensitive Quantitation
The resDNASEQ™ Quantitative CHO DNA System provides highly sensitive detection of CHO DNA, allowing you to use small sample volumes to generate accurate results. The broad linear range of TaqMan™ technology allows you to test samples with variable levels of CHO DNA in the same assay, such as in-process samples with higher amounts of DNA or bulk drug substance with very low amounts. Figure 1 demonstrates the range and sensitivity of the assay. Linearity is demonstrated by analysis of CHO cell standard DNA ranging from 0.3 ng to 3 fg.

Easy to Use, Results in Under 5 Hours
First, you extract DNA from your in-process purification and drug substance samples. Then you perform qPCR to compare DNA amounts in your test samples to a standard curve generated with known amounts of purified CHO cell standard DNA. The easy workflow of the resDNASEQ™ Quantitative CHO DNA System consists of sample preparation, assay setup, and instrument run and data analysis—all of which you can perform in under 5 hours.

Specific to CHO DNA, No Cross-Reactivity With Unrelated DNA
The target of the assay is highly specific, so that it only detects a hamster-specific region of a multicopy genetic element. We selected this region using extensive bioinformatic analysis of multiple related and unrelated species. Testing of assay performance confirmed that the assay is specific to hamster DNA and is unaffected by the presence of as much as 100 ng of unrelated DNA in a test sample.

Consistent Performance Even With Fragmented DNA
For accurate quantitation of residual CHO DNA, assay results must be unaffected by the size of the DNA molecules present in the test sample. To test the effect of DNA fragment size on assay performance, we fragmented high molecular weight CHO genomic DNA into low molecular weight DNA by sonication. Figure 2 demonstrates that the threshold cycle (Ct) values for the reactions with the sonicated low molecular weight DNA were comparable to those of the undigested high molecular weight DNA. These results demonstrate that consistent performance was obtained with the kit irrespective of DNA molecular weight.

TRUSTED_SUSTAINABILITY

Specifica

Content And Storage resDNASEQ CHO DNA Control
• CHO DNA Control, 30 ng/μL, 40 μL, store at –15 to –25°C
• DNA Dilution Buffer (DDB), 7 mL, store at –15 to –25°C before first use and 2−8°C after first use

resDNASEQ CHO DNA Real-Time PCR Reagents
• Environmental Master Mix (2X), 2 x 0.75 mL, store at –15 to –25°C before first use and 2−8°C after first use, protect from light
• CHO DNA Assay Mix (10X), 300 μL, store at –15 to –25°C, protect from light
• Negative Control (water), 1.0 mL, store at –15 to –25°C before first use and 2−8°C after first use

PrepSEQ Nucleic Acid Extraction Kit
• Lysis Buffer (2 bottles, 50 mL/bottle)
• Binding Solution/Isopropanol (1 empty bottle)
• Wash Buffer Concentrate (2 bottles, 26 mL/bottle)
• Elution Buffer (1 bottle, 25 mL)
• Proteinase K Buffer (1 bottle, 50 mL)
• Magnetic Particles (2 tubes, 1.5 mL/tube)
• Proteinase K (20 mg/mL) (1 tube, 1.25 mL), store at or below –20°C
• PK Buffer II (1 bottle, 11 mL)
Store all at room temperature unless otherwise indicated.

PrepSEQ Residual DNA Sample Preparation Kit
• Proteinase K (20 mg/mL), 1 tube, 1.25 mL
• Yeast tRNA (10 mg/mL), 1 tube, 0.5 mL
• Glycogen (5 mg/mL), 2 tubes, 1.0 mL/tube
Store all at or below –20°C.

Metodo di rilevamento Primer-probe
Formato Kit
PCR Method qPCR
Host Cell CHO
Label or Dye FAM
Linea di prodotti resDNASEQ
Quantità 100 reactions
N. di reazioni 100 Reactions
Include Assay Kit and Sample Preparation
compliance-icons compliance-icons
Identificativo prodotto
  • CHO QUANT ASSY+PREPSEQ+MOD.
Avvertenza
  • Pericolo
Categoria di pericolo
  • Tossicità acuta Categoria 4
  • Lesione oculare grave/irritazione oculare Categoria 2
  • Sensibilizzante delle vie respiratorie Categoria 1
  • Corrosione/irritazione della pelle Categoria 2
  • Tossicità specifica per organi bersaglio (esposizione ripetuta) Categoria 2
Indicazioni di pericolo
  • H315-Provoca irritazione cutanea.
  • H319-Provoca grave irritazione oculare.
  • H332-Nocivo se inalato.
  • H334-Può provocare sintomi allergici o asmatici o difficoltà respiratorie se inalato.
  • H373-Può provocare danni agli organi.
Consigli di prudenza
  • P261-Evitare di respirare la polvere/i fumi/i gas/la nebbia/i vapori/gli aerosol.
  • P264-Lavare accuratamente dopo l'uso.
  • P271-Utilizzare soltanto all'aperto o in luogo ben ventilato.
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P284-[Quando la ventilazione del locale è insufficiente] indossare un apparecchio di protezione respiratoria.
  • P302+P352-IN CASO DI CONTATTO CON LA PELLE: lavare abbondantemente con acqua/.
  • P304+P340-IN CASO DI INALAZIONE: trasportare l'infortunato all'aria aperta e mantenerlo a riposo in posizione che favorisca la respirazione.
  • P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
  • P312-Contattare un CENTRO ANTIVELENI/un medico/in caso di malessere.
  • P332+P313-In caso di irritazione della pelle: consultare un medico.
  • P337+P313-Se l'irritazione degli occhi persiste, consultare un medico.
  • P342+P311-In caso di sintomi respiratori: contattare un CENTRO ANTIVELENI/un medico/ .
  • P362+P364-Togliere tutti gli indumenti contaminati e lavarli prima di indossarli nuovamente.
  • P501b-Smaltire il prodotto/recipiente in conformità con le disposizioni locali/regionali/nazionali/internazionali.
Informazioni supplementari
  • MIXTURE LIST-Contain: N-LAUROYLSARCOSINE SODIUM SALT, Thiocyanic acid, Glycerin, Proteinase, tritirachium album serine
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.