Learn More
Thermo Scientific™ Reservoir Accessories
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Tipo:
Sterile Vent Filter
Vent Filter/CO2 Adsorber and Sterile Overflow
Da utilizzare con (apparecchiatura):
Reservoir, 1100 to 2000 L
Storage Tank
Storage Tank 30 to 100 L
Dimensione della confezione:
Pezzo
Descrizione
Level control and sterile overflow for storage reservoirs are perfect accessories for running reverse osmosis systems.
Level control and sterile overflow, Thermo Scientific™ Reservoir Accessories, are perfect for running reverse osmosis systems.
Level Control for Storage Reservoirs
- Required accessory for running any reverse osmosis system; will be adapted to the specific reservoir during the order procedure
Sterile Overflow for Storage Reservoirs
- Recommended accessory to prevent organic contamination inside of reverse osmosis reservoirs
Vent filter for Storage Reservoirs
- Recommended accessory to prevent organic contamination inside of reverse osmosis reservoirs
Vent filter with CO2 absorber for Storage Reservoirs
- Recommended accessory to prevent organic and CO2 contamination inside of reverse osmosis reservoirs
Ordering Information:
This product is not available in all regions
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Reservoir, 1100 to 2000 L |
| Tipo | Vent Filter/CO2 Adsorber and Sterile Overflow |