missing translation for 'onlineSavingsMsg'
Learn More

Synaptobrevin homolog YKT6 Antibody - Azide and BSA Free, R&D Systems™

Product Code. 18621412 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18621412 0.1 mL 0.1mL
18613592 0.02 mL 0.02mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18621412 Supplier Novus Biologicals Supplier No. NBP2948460.1ml
 Restricted Item
Spedizione stimata: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

Synaptobrevin homolog YKT6 Polyclonal specifically detects Synaptobrevin homolog YKT6 in Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptobrevin homolog YKT6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias EC 2.3.1, EC 2.3.1.-, R-SNARE, S. cerevisiae, homolog of, SNARE protein Ykt6, synaptobrevin homolog YKT6, YKT6 v-SNARE homolog (S. cerevisiae), YKT6 v-SNARE protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-198 of human Synaptobrevin homolog YKT6 (NP_006546.1). EFSKQVDRIDWPVGSPATIHYPALDGHLSRYQNPREADPMTKVQAELDETKIILHNTMESLLERGEKLDDLVSKSEVLGTQSKAFYKTARKQNSCCAIM
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10652
Target Species Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.