missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Retinol Binding Protein RBP Antibody (G4E4) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-45017
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Retinol Binding Protein RBP Monoclonal antibody specifically detects Retinol Binding Protein RBP in Human, Mouse, Rat, Goat, Chimpanzee, Monkey, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifica
| Retinol Binding Protein RBP | |
| Monoclonal | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Goat, Chimpanzee | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| G4E4 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 0.5 - 1.0 μg/mL | |
| C, Cellular retinol-binding protein, CRABP-I, CRBP1CRBP-I, CRBPCellular retinol-binding protein I, CRBPI, RBPC, retinol binding protein 1, cellular, retinol-binding protein 1, retinol-binding protein 1, cellular | |
| Retinol binding protein-1 Purified from human plasma | |
| 7 mL | |
| Cancer, Diabetes Research, Lipid and Metabolism | |
| 5947 | |
| Store at 4°C. | |
| IgG1 κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto