Learn More
- Home
- Products
- Specialty Lab Equipment, Instruments, and Apparatuses
- Mills
- RETSCH ZM200 Ultra Centrifugal Mill: Ring Sieves
RETSCH ZM200 Ultra Centrifugal Mill: Ring Sieves
Codice prodotto. 10498722 Sfoglia Tutto Retsch Prodotti
Cambia vista
Click to view available options
Materiale:
Acciaio inox
Stainless Steel
Wear-resistant Stainless Steel
Dimensione del poro:
0.003 in. (0.08 mm)
0.01 in. (0.50 mm)
0.01 in. (0.50 mm)
0.029 in. (0.75 mm)
0.03 in. (1.0 mm)
0.07 in. (2.0 mm)
0.08 mm
0.11 in. (3.0 mm)
0.12 mm
0.20 mm
0.23 in. (6.0 mm)
0.25 mm
0.75 mm
1.0 mm
1.50 mm
2.00 mm
2.0 mm
3.0 mm
3.00 mm
4.0 mm
5.0 mm
5.00 mm
6.0 mm
Stile pori:
Fori trapezoidali
Foro Conidur
Foro trapezoidale
Round holes
Product Type:
Ring Sieve with Reinforced Rim
Setaccio ad anello
Dimensione della confezione:
Pezzo
33 product options available for selection
| Codice prodotto. | Materiale | Dimensione del poro | Stile pori | Product Type | unitSize |
|---|---|---|---|---|---|
| 10498722 | Acciaio inox | 0.01 in. (0.50 mm) | Foro Conidur | Setaccio ad anello | Pezzo |
| 11944242 | Stainless Steel | 1.50 mm | Foro trapezoidale | Ring Sieve with Reinforced Rim | Pezzo |
| 10354721 | Stainless Steel | 1.0 mm | Foro trapezoidale | Setaccio ad anello | Pezzo |
| 10463032 | Acciaio inox | 0.01 in. (0.50 mm) | Foro Conidur | Ring Sieve with Reinforced Rim | Pezzo |
| 10519141 | Acciaio inox | 0.12 mm | Fori trapezoidali | Setaccio ad anello | Pezzo |
| 10670252 | Stainless Steel | 0.20 mm | Foro trapezoidale | Setaccio ad anello | Pezzo |
| 10350061 | Stainless Steel | 0.08 mm | Foro trapezoidale | Setaccio ad anello | Pezzo |
| 10655271 | Stainless Steel | 0.75 mm | Foro trapezoidale | Setaccio ad anello | Pezzo |
| 10130831 | Acciaio inox | 2.00 mm | Fori trapezoidali | Setaccio ad anello | Pezzo |
| 11737704 | Stainless Steel | 4.0 mm | Round holes | Setaccio ad anello | Pezzo |
| 10019060 | Stainless Steel | 1.50 mm | Foro trapezoidale | Setaccio ad anello | Pezzo |
| 10079340 | Acciaio inox | 0.25 mm | Foro trapezoidale | Setaccio ad anello | Pezzo |
| 10730661 | Acciaio inox | 3.00 mm | Round holes | Setaccio ad anello | Pezzo |
| 10039070 | Stainless Steel | 5.0 mm | Round holes | Setaccio ad anello | Pezzo |
| 11761846 | Acciaio inox | 0.029 in. (0.75 mm) | Foro Conidur | Ring Sieve with Reinforced Rim | Pezzo |
| 11904242 | Acciaio inox | 0.003 in. (0.08 mm) | Foro Conidur | Ring Sieve with Reinforced Rim | Pezzo |
| 10297790 | Acciaio inox | 6.0 mm | Round holes | Setaccio ad anello | Pezzo |
| 11914242 | Stainless Steel | 0.12 mm | Fori trapezoidali | Ring Sieve with Reinforced Rim | Pezzo |
| 11924242 | Stainless Steel | 0.20 mm | Foro trapezoidale | Ring Sieve with Reinforced Rim | Pezzo |
| 11954242 | Acciaio inox | 0.07 in. (2.0 mm) | Foro Conidur | Ring Sieve with Reinforced Rim | Pezzo |
| 10101543 | Acciaio inox | 0.03 in. (1.0 mm) | Foro Conidur | Ring Sieve with Reinforced Rim | Pezzo |
| 11974242 | Acciaio inox | 0.11 in. (3.0 mm) | Round holes | Ring Sieve with Reinforced Rim | Pezzo |
| 11964242 | Stainless Steel | 4.0 mm | Round holes | Ring Sieve with Reinforced Rim | Pezzo |
| 11994242 | Acciaio inox | 0.23 in. (6.0 mm) | Round holes | Ring Sieve with Reinforced Rim | Pezzo |
| 11984242 | Stainless Steel | 5.00 mm | Round holes | Ring Sieve with Reinforced Rim | Pezzo |
| 10656320 | Acciaio inox | 0.01 in. (0.50 mm) | Foro Conidur | Ring Sieve with Reinforced Rim | Pezzo |
| 11944252 | Wear-resistant Stainless Steel | 0.25 mm | Foro trapezoidale | Ring Sieve with Reinforced Rim | Pezzo |
| 11702513 | Wear-resistant Stainless Steel | 0.20 mm | Foro trapezoidale | Ring Sieve with Reinforced Rim | Pezzo |
| 11954252 | Wear-resistant Stainless Steel | 0.75 mm | Foro trapezoidale | Ring Sieve with Reinforced Rim | Pezzo |
| 11984252 | Wear-resistant Stainless Steel | 2.0 mm | Foro trapezoidale | Ring Sieve with Reinforced Rim | Pezzo |
| 11846013 | Wear-resistant Stainless Steel | 1.0 mm | Foro trapezoidale | Ring Sieve with Reinforced Rim | Pezzo |
| 11994252 | Wear-resistant Stainless Steel | 3.0 mm | Round holes | Ring Sieve with Reinforced Rim | Pezzo |
| 11974252 | Wear-resistant Stainless Steel | 1.50 mm | Foro trapezoidale | Ring Sieve with Reinforced Rim | Pezzo |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 33 options available.
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Codice prodotto.
10498722
Fornitore
RETSCH
N. del fornitore
03.647.0235
Accedi o Registrati per verificare il tuo prezzo e la disponibilità.
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Specifica
Specifica
| Product Type | Setaccio ad anello |
| Da utilizzare con (apparecchiatura) | Mulino centrifugo ZM200 Ultra |
| Materiale | Acciaio inox |
| Dimensione del poro | 0.01 in. (0.50 mm) |
| Stile pori | Foro Conidur |