Learn More
3M™ Reusable Half Mask Respirator Kit A2P3 R
Click to view available options
Size:
Medium
Large
Dimensione della confezione:
Pezzo
Descrizione
- Ready to use respiratory kit with 3M™ Reusable Half Face Mask 7503L or 7503M and 3M™ Filters offering A2P3 R protection
- Half mask with a soft silicone face piece for comfort
- Bayonet connection fitting system to easily connect filters to mask
- 3M™ Cool Flow™ Valve provides easier breathing and reduced heat and moisture build-up
- Low profile, twin filter design half mask for a balanced secure fit
Specifica
Specifica
| Certificazione/Conformità | CE Approved |
| Tipo di filtro | A2P3 R |
| Product Type | Half Mask Respirator Kit |
| Size | Medium |
| Include | Half mask and filter |