Learn More
R&D Systems™ Rhesus EDA2R/TNFRSF27/XEDAR (NP_001180396.1) VersaClone cDNA
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
Specifica
Specifica
| Gene | EDA2R/TNFRSF27/XEDAR |
| Numero di accesso | NP_001180396.1 |
| ID gene (immissione) | 60401 |
| Alias gene | ectodysplasin A2 receptor, EDA-A2 receptor, EDAA2R, EDA-A2R, EDAA2Rtumor necrosis factor receptor superfamily member XEDAR, EDAR2, TNFRSF27, XEDAR, XEDARTNFRSF27tumor necrosis factor receptor superfamily member 27, X-linked ectodysplasin-A2 receptor |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Specie | Rhesus Macaque |
| Quantità | 10 μg |