Learn More
Ribosomal Protein S6/RPS6 Antibody (522731), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals FAB5436MFV500
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Ribosomal Protein S6/RPS6 Monoclonal antibody specifically detects Ribosomal Protein S6/RPS6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry
Specifica
| Ribosomal Protein S6/RPS6 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 522731 | |
| Western Blot, Immunocytochemistry | |
| Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, S6 | |
| E. coli-derived recombinant human RPS6, Met1-Lys249, Accession # P62753 | |
| 0.1 mL | |
| Cancer, DNA Repair, mTOR Pathway, Phospho Specific, Translation Control | |
| 6194 | |
| Store at 4C in the dark. | |
| IgG2b |