Learn More
Epredia™ Dry Specimen Containers - Snap Lid
Sfoglia Tutto Epredia Prodotti
Click to view available options
Capacità (sistema metrico):
474 mL
Dimensione della confezione:
pezzi 100
Descrizione
Three unique packaging configurations allow for easy distribution and transport.
53 Series Containers
- Easy to transport trays ideal for operating rooms and hospitals
- Clear polystyrene containers have screw-top lids
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
56 Series Containers
- Patented Epredia™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
- Easy to transport trays
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
59 Series Containers
- Patented Epredia Bio-Tite closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
- Stack pack reduces excess packaging
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Specifica
Specifica
| Product Type | Dry Specimen Container |
| Closure Included | Yes |
| Materiale | Polyethylene |
| Sterilità | Non-sterile |
| Capacità (sistema metrico) | 474 mL |
| Tipo di chiusura | Snap Cap |
| Colore | Clear |
| Descrizione | Dry Containers |
| N. per custodia | 100/Cs. |