Learn More
Epredia™ Richard-Allan Scientific™ Modified Wright Stain Pack
Sfoglia Tutto Epredia Prodotti
Click to view available options
Tipo:
Modified Wright Stain Pack
Dimensione della confezione:
pezzi 5
Descrizione
This system consists of a modified polychrome methylene blue-eosin stain (similar to Wright or Giemsa) and based on the original stain proposed by Romanowsky*.
Specifica
Specifica
| Quantità | 5 Pk. |
| Tipo | Modified Wright Stain Pack |