Learn More
Epredia™ Signature Series™ Pen-Fix™
Codice prodotto. p-4532389 Sfoglia Tutto Epredia Prodotti
Click to view available options
Quantità:
4 x 1 gal.
Dimensione della confezione:
pezzi 4
Codice prodotto. 10148438
Marca: Epredia™ 6101
Achieve fixation and dehydration of specimens simultaneously with Epredia™ Signature Series™ Pen-Fix™, a uniquely buffered alcoholic formalin fixative.
  • Excellent frozen section fixative
  • Performs well with fatty or thick tissue sections
  • Penetration rate three times faster than 10% Neutral Buffered Formalin
  • Can be used on all open or enclosed tissue processors and in the grossing room
  • Recommended for use in the second and/or third stations of the processor as a secondary fixative
TRUSTED_SUSTAINABILITY
Specifica
Quantità 4 x 1 gal.