Learn More
Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver, Silver Nitrate #2 Solution
Codice prodotto. 10485709 Sfoglia Tutto Epredia Prodotti
Click to view available options
Tipo:
Gum Mastic Solution
Hydroquinone
Sensitizer Solution
Silver Nitrate Solution #1
Steiner-Steiner Silver Kit
Dimensione della confezione:
500mL
Pezzo
pezzi 12
Codice prodotto. 10485709
Marca: Epredia™ 87009
Determine the presence of spirochetes and nonfilamentous bacteria with Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver.
  • Kit does not contain uranyl nitrate
  • Background is stained yellow to brown
  • Bacteria, Donovan bodies and fungi all stain black
  • Kit formulation and detailed procedures provide consistent and reproducible results
TRUSTED_SUSTAINABILITY
Specifica
Tipo Steiner-Steiner Silver Kit