Learn More
Quickfit™ Still Head Vertical Recovery Adapter
Descrizione
Used in distillation assemblies in which the condenser is assembled in the vertical mode
- Used in distillation assemblies in which the condenser is assembled in the vertical mode
Specifica
Specifica
| Da utilizzare con (applicazione) | Used in distillation assemblies in which the condenser is assembled in the vertical mode |
| Tipo | Vertical Recovery Still Head |
| Materiale | Vetro borosilicato |
| Certificazione/Conformità | ISO 3585 |
| Size | 19/26 (Cone) |