Learn More
Corning™ rLaminin-521 (Human)
Codice prodotto. 15689176 Sfoglia Tutto Corning Prodotti
Click to view available options
Quantità:
1 mg
100 μg
5 mg
Dimensione della confezione:
100µg
5mg
Pezzo
Codice prodotto. 15689176
Marca: Corning™ 354221
Recombinant heterotrimer protein composed of α5, β2, and γ1 chains
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning rLaminin-521 is the next generation pluripotent stem cell (PSC) surface that enables researchers to streamline culture workflow, lower contamination risk, and scale-up efficiently. Corning has partnered with BioLamina for the supply of recombinant human Laminin 521 (rLaminin-521), a biocompatible, full length Laminin that enables feeder-free, clump-free, single-cell passaging of PSCs without ROCK-inhibitor.
TRUSTED_SUSTAINABILITY
Specifica
Nome rLaminin-521, Human
Quantità 100 μg
Sorgente Human
Recombinant Recombinant