Learn More
RNASEL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 22577-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
A virally induced endoribonuclease, RNase L is the terminal factor of the anti-viral action of interferon. Activation of RNase L results in inhibition of viral proliferation. Activation of RNase L has also been determined to degrade RNA, thus, initiating a cellular stress response, inducing apoptosis. Since mutations in RNase L relate to an increased incidence of prostate cancer, the activation of RNase L functions in counteracting prostate cancer, via apoptosis.Specifica
| RNASEL | |
| Polyclonal | |
| Unconjugated | |
| RNASEL | |
| 2 5A dependent ribonuclease, 2 5A dependent RNase, PRCA1, Ribonuclease 4, Ribonuclease L, RNase L, RNASEL, RNS4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24014, 359726, 6041 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q05823, Q05921 | |
| Rnasel | |
| RNASEL Fusion Protein Ag17409 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |