Learn More
Bel-Art™ Robotic Sterile Pipette Tips
Sfoglia Tutto SP Scienceware Prodotti
Click to view available options
Da utilizzare con (applicazione):
For: Hamilton, Volume: 100 μL, Conductive, Material: carbon filled polypropylene
For: Hamilton, Volume: 100 μL, Non-conductive, Material: Clear polypropylene
For: Hamilton, Volume: 50 μL, Conductive, Material: carbon filled polypropylene
For: Hamilton, Volume: 50 μL, Non-conductive, Material: clear polypropylene
For: Tecan, Volume: 100 μL, Conductive, Material: carbon filled polypropylene
For: Tecan, Volume: 100 μL, Non-conductive, Material: clear polypropylene
For: Tecan, Volume: 50 μL, Conductive, Material: carbon filled polypropylene
For: Tecan, Volume: 50 μL, Non-conductive, Material: clear polypropylene
Dimensione della confezione:
pezzi 16
pezzi 24
Descrizione
- Sterile, disposable, filtered pipette tips can be used in robotic systems
- Filtered to prevent contamination of pipette arm
- Conductive tip models detect liquid level when pipetting conductive liquids
- Ensures repeatable volume accuracy
- DNase, RNase, pyrogen free, manufactured in Class 8 clean room
- CE marked
Specifica
Specifica
| Volume (sistema metrico) | 1000 μL |
| Filtrato | Con filtro |
| Sterilità | Sterile |
| Colore | Trasparente |
| Autoclavabile | Not Autoclavable |
| Certificazione/Conformità | CE marked |
| Da utilizzare con (applicazione) | For: Tecan, Volume: 100 μL, Non-conductive, Material: clear polypropylene |
| Formato | Hanging Tip Rack |
| Stile punta | Standard |
| Imballaggio | 16 racks x 96 tips |
| Vedi altri risultati |