Learn More
ROR1 Mouse anti-Human, Unconjugated, Clone: 1F5D8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66923-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The ROR1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ROR1. This antibody recognizes human antigen. The ROR1 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifica
| ROR1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA | |
| 1F5D8 | |
| Unconjugated | |
| 150 μL | |
| 4919 | |
| IgG1 |