Learn More
Quickfit™ Plain Side-Arms
Codice prodotto. p-8002704
Click to view available options
Diametro (sistema metrico):
8 mm
10 mm
13 mm
Dimensione della confezione:
pezzi 10
pezzi 5
Codice prodotto. 12485266
Marca: Quickfit GP3RA/5
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Design gives excellent flow control and allows use with fluids and gases having temperatures between -20°C and 200°C.
  • Rotary design for fine single drop control invaluable on burettes
  • Borosilicate glass with PTFE key for excellent chemical resistance
  • No lubrication required - no seizure and no contamination
  • Temperature range -20°C to 200°C
  • Vacuum capability to 10-4Torr
  • Increased barrel diameter for excellent drainage
  • Locking ring to prevent accidental removal
  • Ergonomic key for good grip and easy operation
TRUSTED_SUSTAINABILITY
Specifica
Tipo Plain Side-arm
Diametro (sistema metrico) 8 mm
Materiale Borosilicate Glass, PTFE (Key)
Da utilizzare con (apparecchiatura) GP Rotaflo™ stopcock