Learn More
Rotaflo™ Straight Pattern Plain-side Arms for Burettes
Click to view available options
Diametro (sistema metrico):
8 mm
10 mm
13 mm
Dimensione della confezione:
pezzi 10
pezzi 5
Descrizione
- No lubrication required - no seizure and no contamination
- Temperature range: -20°C to 200°C
- Vacuum capability to 10 Torr
- Increased barrel diameter for drainage
- Locking ring to prevent accidental removal
- Ergonomic key for good grip and easy operation
Specifica
Specifica
| Tipo | Plain Side-arm |
| Diametro (sistema metrico) | 10 mm |
| Materiale | Borosilicate Glass |
| Da utilizzare con (apparecchiatura) | GP Rotaflo™ Stopcock |