Learn More
BUCHI Rotavapor I-300 Vacuum Controller and Interface
Sfoglia Tutto Buchi Prodotti
Click to view available options
Modello:
Pro
Standard
Dimensione della confezione:
Pezzo
Descrizione
- Easy plug and play integration into a Rotavapor R-300 system at any time
- Central control of all process parameters such as rotation speed, temperature (heating, cooling, vapor) and pressure
- Remote monitoring on smart phones including push notifications and live view
- Modes: manual, timer, continuous
- Flexibility of the interface allows easy installation and convenient operation from multiple positions:
- Rotavapor handle
- Vacuum pump
- Fume hood or lab bench wall
- Stand-alone operation to locally control a central vacuum system
- Synchronized processing thanks to centrally controlled rotary evaporator, heating bath, speed-controlled vacuum pump and recirculating chiller
- Unattended operation (optional):
- Automatic distillation with CloudDest sensor
- Foam sensor for foaming samples
Specifica
Specifica
| Tipo | Vacuum Tubing |
| Include | Kit for R-21x/V-7x0 |
| Modello | Standard |