Learn More
BUCHI Rotavapor I-300 Vacuum Controller and Interface
Codice prodotto. p-200136244 Sfoglia Tutto Buchi Prodotti
Click to view available options
Modello:
Pro
Standard
Dimensione della confezione:
Pezzo
Codice prodotto. 17274694
Marca: BUCHI 11I300P40
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
The BUCHI Interface I-300 controls all distillation parameters of a Rotavapor R-300 system including the optional V-300 Vacuum Pump and F-3XX series chillers.
  • Easy plug and play integration into a Rotavapor R-300 system at any time
  • Central control of all process parameters such as rotation speed, temperature (heating, cooling, vapor) and pressure
  • Remote monitoring on smart phones including push notifications and live view
  • Modes: manual, timer, continuous
  • Flexibility of the interface allows easy installation and convenient operation from multiple positions:
    • Rotavapor handle
    • Vacuum pump
    • Fume hood or lab bench wall
  • Stand-alone operation to locally control a central vacuum system
  • Synchronized processing thanks to centrally controlled rotary evaporator, heating bath, speed-controlled vacuum pump and recirculating chiller
  • Unattended operation (optional):
    • Automatic distillation with CloudDest sensor
    • Foam sensor for foaming samples
Specifica
Tipo Vacuum Tubing
Include Kit for R-21x/V-7x0
Modello Standard