Learn More
OHAUS™ Rotor Swing Out ID Sealable
Sfoglia Tutto Ohaus Prodotti
Click to view available options
Capacità:
4 x 750 mL
Dimensione della confezione:
Pezzo
Descrizione
- Choose from a range of rotors to be used with Frontier centrifuges to match capacities and flexibility required for your basic to high-performance sample preparation needs.
- Designed with usability in mind, the Frontier rotors can be effortlessly installed or removed in a few quick and simple steps, and users have easy access to the samples.
- The rotors are durably constructed of high-quality aluminum alloy or polypropylene to last through demanding workflows - from autoclave sterilization to sub-zero temperatures.
Specifica
Specifica
| Capacità | 4 x 750 mL |
| Velocità max | 4,500,000 rpm |