Learn More
SVL™ Rotulex™ Spherical Joints
Click to view available options
N. giunto rastremato standard:
13
19
29
35
41
64
Include:
Female Joint
Female Joint, PTFE Covered O-ring, Spring-loaded Metal clip
Male Joint
Male Joint, PTFE Covered O-ring, Spring-loaded Metal clip
Dimensione della confezione:
pezzi 20
pezzi 30
pezzi 5
Descrizione
- All glass and PTFE pathway is highly resistant to chemical attack
- Can be used at temperatures up to 200°C
- System comprises male ball joint with PTFE covered O-ring which fits into female cup joint
- Joint can be retained together with spring loaded metal clip
- Component parts RM, RF, RO and RC should be ordered separately
Specifica
Specifica
| Include | Female Joint |
| Materiale | Glass, PTFE |
| Product Type | Spherical joint |
| Quantità | 5/Pk. |
| N. giunto rastremato standard | 64 |