Learn More
Epredia™ Digital Round Tissue Section Bath
Codice prodotto. p-6899008 Sfoglia Tutto Epredia Prodotti
Click to view available options
Tensione:
230 V
Dimensione della confezione:
Pezzo
Codice prodotto. 15553646
Marca: Epredia™ A84600060
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Obtain accuracy, reliability, and ease-of-use with the Epredia™ Digital Round Tissue Section Bath.
Features
  • Deep bowl for easier section pick up
  • Wide bowl accommodates longer section ribbons
  • Large area to rest hands when floating sections
  • Miniature circuit breakers for ease-of-use; simply push to reset
  • Anti-microbial paint finish (silver ion) to inhibit bacterial growth
Heating Efficiency
  • Ultra-fast heating system
  • Significant slide drying area at the rear of the instrument
  • A secure-fitting lid reduces airborne contamination, evaporation, and improves heating efficiency
  • Automatic over-temperature cut-out for safety
TRUSTED_SUSTAINABILITY
Specifica
Capacità (sistema metrico) 3.5 L
Certificazione/Conformità Certificate of Analysis (COA), Certificate of Conformance (COC)
Comunicazioni Digital
Tensione 230 V