missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15387-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This gene encodes a protein component of the 60S ribosomal subunit. This protein can bind specifically to domain C of the tax-responsive enhancer element of human T-cell leukemia virus type 1, and may participate in tax-mediated transactivation of transcription. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed throughout the genome. Alternative splicing results in multiple transcript variants.Specifica
| RPL6 | |
| Polyclonal | |
| Unconjugated | |
| RPL6 | |
| 60S ribosomal protein L6, Neoplasm related protein C140, ribosomal protein L6, RPL6, SHUJUN 2, TAXREB107, TXREB1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117042, 19988, 6128 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P21533, P47911, Q02878 | |
| RPL6 | |
| RPL6 Fusion Protein Ag6840 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto