Learn More
Invitrogen™ RSPO1 Monoclonal Antibody (7A6)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA515489
Beschreibung
MA5-15489 targets R-spondin1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15489 immunogen is purified recombinant fragment of R-spondin1 expressed in E. Coli.. MA5-15489 detects R-spondin1 which has a predicted molecular weight of approximately 28.9kDa.
This gene is a member of the R-spondin family and encodes a secreted activator protein with two cystein-rich, furin-like domains and one thrombospondin type 1 domain. In mice, the protein induces the rapid onset of crypt cell proliferation and increases intestinal epithelial healing, providing a protective effect against chemotherapy-induced adverse effects.
Spezifikation
| RSPO1 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| Q2MKA7 | |
| RSPO1 | |
| Purified recombinant fragment of R-spondin1 expressed in E. Coli. | |
| RUO | |
| 284654 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 7A6 | |
| Unconjugated | |
| RSPO1 | |
| CRISTIN3; cristin-3; cysteine-rich and single thrombospondin domain-containing protein 3; FLJ40906; hRspo1; mCristin-3; RGD1565558; Roof plate-specific spondin-1; RP11-566C13.1; RSPO; RSPO1; Rspondin; R-spondin; R-spondin 1; R-spondin homolog; R-spondin homolog (Xenopus laevis); R-spondin-1; thrombospondin type 1 domain containing gene Rspondin | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.