missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSPO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA520749
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
A suggested positive control is rat spleen tissue lysate. PA5-20749 can be used with blocking peptide PEP-0863.
This gene is a member of the R-spondin family and encodes a secreted activator protein with two cystein-rich, furin-like domains and one thrombospondin type 1 domain. In mice, the protein induces the rapid onset of crypt cell proliferation and increases intestinal epithelial healing, providing a protective effect against chemotherapy-induced adverse effects.
Specifica
| RSPO1 | |
| Polyclonal | |
| Unconjugated | |
| RSPO1 | |
| CRISTIN3; cristin-3; cysteine-rich and single thrombospondin domain-containing protein 3; FLJ40906; hRspo1; mCristin-3; RGD1565558; Roof plate-specific spondin-1; RP11-566C13.1; RSPO; RSPO1; Rspondin; R-spondin; R-spondin 1; R-spondin homolog; R-spondin homolog (Xenopus laevis); R-spondin-1; thrombospondin type 1 domain containing gene Rspondin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 192199, 284654 | |
| 4°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q2MKA7, Q9Z132 | |
| RSPO1 | |
| A 16 amino acid peptide from near the amino terminus of human RSPO1. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto