missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSPO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA534937
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Recommended positive controls: HL-60. Predicted reactivity: Mouse (90%), Rat (87%), Dog (91%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of the R-spondin family and encodes a secreted activator protein with two cystein-rich, furin-like domains and one thrombospondin type 1 domain. In mice, the protein induces the rapid onset of crypt cell proliferation and increases intestinal epithelial healing, providing a protective effect against chemotherapy-induced adverse effects.
Specifica
| RSPO1 | |
| Polyclonal | |
| Unconjugated | |
| RSPO1 | |
| CRISTIN3; cristin-3; cysteine-rich and single thrombospondin domain-containing protein 3; FLJ40906; hRspo1; mCristin-3; RGD1565558; Roof plate-specific spondin-1; RP11-566C13.1; RSPO; RSPO1; Rspondin; R-spondin; R-spondin 1; R-spondin homolog; R-spondin homolog (Xenopus laevis); R-spondin-1; thrombospondin type 1 domain containing gene Rspondin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 284654 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q2MKA7 | |
| RSPO1 | |
| Recombinant protein encompassing a sequence within the center region of human RSPO1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto