missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSPO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA583809
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: CKEGLYLHKG RCYPACPEGS SAANGTMECS SPAQCEMSEW SPWGPCSKKQ QLCGFRRGSE ERTRRVLHA Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 90%.
This gene is a member of the R-spondin family and encodes a secreted activator protein with two cystein-rich, furin-like domains and one thrombospondin type 1 domain. In mice, the protein induces the rapid onset of crypt cell proliferation and increases intestinal epithelial healing, providing a protective effect against chemotherapy-induced adverse effects.
Specifica
| RSPO1 | |
| Polyclonal | |
| Unconjugated | |
| RSPO1 | |
| CRISTIN3; cristin-3; cysteine-rich and single thrombospondin domain-containing protein 3; FLJ40906; hRspo1; mCristin-3; RGD1565558; Roof plate-specific spondin-1; RP11-566C13.1; RSPO; RSPO1; Rspondin; R-spondin; R-spondin 1; R-spondin homolog; R-spondin homolog (Xenopus laevis); R-spondin-1; thrombospondin type 1 domain containing gene Rspondin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 284654 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q2MKA7 | |
| RSPO1 | |
| Recombinant protein corresponding to Human RSPO1. Recombinant protein control fragment (Product #RP-94446). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto