missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSPO3 Mouse anti-Human, Clone: 1E4F1, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66314-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
RSPO3 is a member of the R-spondin family and plays a role in the regulation of Wnt (wingless-type MMTV integration site family)/beta-catenin and Wnt/planar cell polarity (PCP) signaling pathways, which are involved in development, cell growth and disease pathogenesis. RSPO3 has been shown to be involved in regulating the balance between angioblast and blood cell specification during embryonic vasculogenesis and angiogenesis and may be involved in tumor development.Specifica
| RSPO3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BXY4 | |
| RSPO3 | |
| RSPO3 Fusion Protein Ag9899 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1E4F1 | |
| Unconjugated | |
| RSPO3 | |
| CRISTIN1, hPWTSR, hRspo3, Protein with TSP type 1 repeat, PWTSR, R spondin 3, Roof plate specific spondin 3, RSPO3, THSD2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 84870 | |
| -20°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto