missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA564190
This item is not returnable.
View return policy
Description
Immunogen sequence: CSWSQYEMPE FSSDSTMRIL SNLVQHILKK YYGNGTRKSP EMPVILEVSL EGSHDTANCE ACTLGICG Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 48%.
RTP4 belongs to the TMEM7 family. The function of this protein remains unknown.Specifications
RTP4 | |
Polyclonal | |
Unconjugated | |
RTP4 | |
28 kDa interferon-responsive protein; 28kD interferon alpha responsive protein; 28kD interferon responsive protein; 3CxxC-type zinc finger protein 4; 5830458K16Rik; IFRG28; receptor (chemosensory) transporter protein 4; receptor transporter protein 4; receptor transporting protein 4; receptor-transporting protein 4; Rtp4; Z3CXXC4; zinc finger, 3CxxC-type 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64108 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96DX8 | |
RTP4 | |
Recombinant protein corresponding to Human RTP4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only