Learn More
Tocris Bioscience™ RU.521
Click to view available options
Quantità:
10 mg
50 mg
Dimensione della confezione:
10mg
50mg
Descrizione
RU.521 is a potent and selective cyclic GMP-AMP synthase (cGAS) inhibitor (Kd = 36.2 nM; IC50 values are 0.11 μM and 2.94 μM against m-cGAS and h-cGAS, respectively). RU.521 suppresses dsDNA-activated reporter activity but not 5'ppp-HP20 RNA or murine IFN-β-induced IFN-β1-dependent gene expression. RU.521 reduces constitutive expression of type I interferon in primary macrophages from a mouse model of Aicardi-Gouti?res syndrome. Pretreatment of chimeric antigen receptor (CAR) redirected T cells with RU.521 increases efficiency of virus-free gene editing by improving cell viability and CAR integration in cell line.
Specifica
Specifica
| Materiale o nome chimico | 3-[1-(6,7-Dichloro-1 H-benzimidazol-2-yl)-5-hydroxy-3-methyl-1 H-pyrazol-4-yl]-1(3 H)-isobenzofuranone |
| CAS | 2262452-06-0 |
| Quantità | 10 mg |
| Bersaglio | Synthase/Synthetase Inhibitors |
| Formula molecolare | C19H12Cl2N4O3 |
| Purezza | 0.98 |