Learn More
Abeba™ Rubber 5150 Shoes
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- Dark blue
- PU midsole
- Slip resistance special rubber outsole sole
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Leather top layer
- Adjustable instep area
- Lining microfiber, respiration absorbing
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20347:2012 OB A, FO SRC |
| Colore | Blue |
| Materiale rivestimento interno | Microfiber |
| Materiale | Leather |
| Size | 46 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Rubber |
| Stile | 5150 |