Learn More
Abeba™ Rubber 5310 Shoes
Descrizione
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather, perforated
- Black
- PU midsole
- Slip resistance special rubber outsole sole
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
- Leather top layer
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20347:2012 OB A, FO SRC |
| Colore | Black |
| Materiale | Leather |
| Size | 44 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Rubber |
| Stile | 5310 |