Learn More
Uvex™ Rubiflex NB 3501 Safety Gloves
Click to view available options
Size:
7
8
9
10
Descrizione
- Full NBR-coating is highly flexible for tasks that require dexterity
- Breathabel RUBIPOR impregnation
- Cotton interlock liner
- Resistant to oil, grease, fuel, and aerosols
- Ideal for use in dry environments
Specifica
Applicazioni | Industrial |
Coating Material | NBR |
Colore | Orange |
Size | 8 |
EN388 Standard | 0121X |
Da utilizzare con (applicazione) | Inspection, Light Metal Work, Maintanence, and Painting |
Sesso | Unisex |
Stile polso guanto | Gauntlet |
Materiale rivestimento interno | Cotton |
Serie | Rubiflex NB 3501 |