Learn More
uvex Rubiflex NB27 Safety Gloves
Sfoglia Tutto Uvex Prodotti
Click to view available options
Size:
9
10
11
Dimensione della confezione:
10 paia
Descrizione
- NBR-coated safety gloves with cotton interlock liner
- Hard-wearing, durable gloves are highly flexible
- Ideal for tough jobs in damp, oily, or greasy work environments
- Free of harmful substances in accordance with OEKO-TEX™ Standard 100
Specifica
Specifica
| Certificazione/Conformità | OEKO-TEX™ Standard 100 |
| Applicazioni | Laboratory |
| Coating Material | NBR |
| Colore | Orange |
| Latex Free | Sì |
| Materiale | Nitrile |
| Product Type | Chemical-resistant Gloves |
| Size | 9 |
| EN388 Standard | 3111X |
| Da utilizzare con (applicazione) | Inspection, Maintenance, Metal Processing, Painting, and Repairs |
| Vedi altri risultati |