Learn More
(S)-2-(Boc-amino)butyric acid, 95%
Codice prodotto. p-7055172
Click to view available options
Quantità:
1 g
5 g
25 g
Dimensione della confezione:
1g
25g
5g
Codice prodotto. 15400509
Marca: Thermo Scientific Alfa Aesar H63686.03
Richiesta di formato massivo o personalizzato
CAS: 34306-42-8 | C9H17NO4 | 203.24 g/mol
It is employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 34306-42-8
Formula molecolare C9H17NO4
Molecular Weight (g/mol) 203.24
Numero MDL MFCD00037267
InChI Key PNFVIPIQXAIUAY-UHFFFAOYNA-N
Sinonimo boc-abu-oh, boc-l-2-aminobutyric acid, boc-2-abu-oh, s-2-tert-butoxycarbonyl amino butanoic acid, boc-l-2-aminobutanoic acid, boc-l-abu-oh, s-2-tert-butoxycarbonylamino butyric acid, s-2-boc-amino butyric acid, 2s-2-tert-butoxycarbonyl amino butanoic acid, l-2-boc-amino butyric acid
PubChem CID 2755934
SMILES CCC(NC(=O)OC(C)(C)C)C(O)=O
Specifica
Materiale o nome chimico (S)-2-(Boc-amino)butyric acid
Quantità 1 g
Beilstein 6801706
Informazioni di solubilità Slightly soluble in water.
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
Peso formulazione 203.24
Percent Purity 95%
Forma fisica Solid