missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100A9 Monoclonal Antibody (01)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA529543
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human S100A9.
S100A9 (MRP-14) is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis.
Specifica
| S100A9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P06702 | |
| S100A9 | |
| Recombinant Human S100A9 protein (Met1-Pro114). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry | |
| 01 | |
| Unconjugated | |
| S100A9 | |
| 60B8AG; AW546964; BEE22; BM-009; Cagb; calgranulin B; calgranulin-B; Calprotectin L1H subunit; CFAG; CGLB; DKFZp686B04128; GAGB; intracellular calcium-binding protein (MRP14); L1AG; Leukocyte L1 complex heavy chain; LIAG; MAC387; Macrophage; MIF; Migration inhibitory factor-related protein 14; Monocyte; MRP14; MRP-14; MRP15; MRP16; myeloid-related protein 14; NIF; P14; protein S100-A9; S100 A9; S100 calcium binding protein A9; S100 calcium binding protein A9 (calgranulin B); S100 calcium-binding protein A9; S100 calcium-binding protein A9 (calgranulin B); S100a9 | |
| Mouse | |
| Protein A | |
| RUO | |
| 6280 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto