Learn More
DWK Life Sciences Safeguarding, for safety standard ground sockets
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
Dimensione della confezione:
Pezzo
pezzi 10
Descrizione
Polyamide tension rings have very good sliding properties. They help to safely loosen fixed joints through even distribution of force.
- By means of an even force distribution, stuck joints can be safely loosened.
- Due to their material properties, pull rings have very good sliding properties.
Specifica
Specifica
| Quantità | 10/Pk. |
| Descrizione articolo | To secure the security grinding |
| Tipo | Safeguard |
| Joint Size | NS 24/29 |