Learn More
Uvex Safety Gloves Phynomic 60068
Sfoglia Tutto Uvex Prodotti
Click to view available options
Size:
6
7
8
9
10
11
12
Dimensione della confezione:
10 paia
Descrizione
- Free of accelerators and dermatologically tested for skin compatibility (proDERM Institute)
- Ideal for allergy sufferers
Specifica
Specifica
| Coating Material | Aqua-Polymer |
| Colore | Black |
| Cut Resistance Rating | Level F |
| Hand | Ambidestri |
| Latex Free | Sì |
| Materiale | Elastane, HPPE, Polyamide, Steel Fiber |
| Product Type | Mechanical/Cut Protection Gloves (Heavy-duty) |
| Size | 12 |