Learn More
Uvex Safety Gloves Phynomic 60068
Codice prodotto. p-200105925 Sfoglia Tutto Uvex Prodotti
Click to view available options
Size:
6
7
8
9
10
11
12
Dimensione della confezione:
10 paia
Codice prodotto. 30245729
Marca: Uvex 6006806
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Phynomic assembly and cut protection gloves are especially ergonomically shaped, sit like a second skin and are very light, flexible and breathable.
  • Free of accelerators and dermatologically tested for skin compatibility (proDERM Institute)
  • Ideal for allergy sufferers
TRUSTED_SUSTAINABILITY
Specifica
Coating Material Aqua-Polymer
Colore Black
Cut Resistance Rating Level F
Hand Ambidestri
Latex Free
Materiale Elastane, HPPE, Polyamide, Steel Fiber
Product Type Mechanical/Cut Protection Gloves (Heavy-duty)
Size 6